Hematoxylin

Supplier: Thermo Scientific Chemicals

41118-0250
TS41118-0250EA 67 USD
TS41118-0250
Hematoxylin
Hematoxylin
Formula: C₁₆H₁₄O₆
Melting Pt: 291…293 °C
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654

Order Now

Specification Test Results

Appearance Beige to orange to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (1 g, 105 °C, 3 hrs)
UV spectrophotometry ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR