Gibberellic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
41091-0010
41091-0050
41091-0250
TS41091-0010EA
68
USD
TS41091-0010
TS41091-0050
TS41091-0250
Gibberellic acid 99%
Gibberellic acid
Formula:
C₁₉H₂₂O₆ Melting Pt: 227 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 EINECS: 201-001-0 Merck Index: 13,04430 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % (K.F.) |
Specific optical rotation | +71° to +81° (20°C, 589 nm) (c=2, methanol) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...