Gallic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid

41086-0010 41086-0050 41086-0051 41086-2500
TS41086-0010EA 408 USD
TS41086-0010 TS41086-0050 TS41086-0051 TS41086-2500
Gallic acid 98%
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
Melting Pt: 253 °C (dec.)
Density: 1.694 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
GC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR