Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
AC41050-0250
AC41050-1000
AC41050-5000
TS41050-0250EA
33
USD
TS41050-0250
TS41050-1000
TS41050-5000
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Formula:
C₃H₅KOS₂ Melting Pt: 209…214 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004931 CAS Number: 140-89-6 EINECS: 205-439-3 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
Titration with HClO4 | ≥97.0 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...