Ethyl oleate mixture of homologeous fatty acid esters 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
41029-0010
41029-0025
41029-2500
TS41029-0010EA
114
USD
TS41029-0010
TS41029-0025
TS41029-2500
Ethyl oleate mixture of homologeous fatty acid esters 98%
Ethyl oleate
Formula:
C₂₀H₃₈O₂ Boiling Pt: 190…210 °C (7 mmHg) Density: 0.86 g/cm³ Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009579 CAS Number: 111-62-6 |
Specification Test Results
Appearance | Clear viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4530 (20°C, 589 nm) |
Assay | ≥97.5 % (ester) |
Water | ≤1 % (K.F.) |
Density | (20°C) 0.869 to 0.874 |
GC | ≥60 % (oleic acid) |
Color scale | ≤70 APHA |
Stabilizer | 80 to 120 ppm BHA content |
Acid value | ≤0.5 mg KOH/g |
Saponification value | 177 to 188 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...