N,N-Dimethylformamide ≥99.8% ACS for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

40832-0010 40832-5000 408320025
TS40832-0010EA 162 USD
TS40832-0010 TS40832-5000 TS408320025
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.945 g/cm³
Flash Pt: 58 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.15 % (Coulometric)
GC ≥99.8 %
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.0005 meq/g
Titrable base ≤0.003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR