Dextran for biochemistry, high fraction
Supplier: Thermo Scientific Chemicals
40626-0010
40626-1000
TS40626-0010EA
726
USD
TS40626-0010
TS40626-1000
Dextran for biochemistry, high fraction
Dextran
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤9.0 % |
Sulfated ash | ≤2.0 % |
UV | at 375 nm A: ≤0.15 |
Viscosity | 0.4 to 0.5 (1 % at 37°C) |
Average molecular weight | 250000 |
pH | 4.5 to 7.0 (6 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...