Cyclohexanone 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pimelic Ketone

406090010 406090025 406092500
TS406090010EA 73 USD
TS406090010 TS406090025 TS406092500
Cyclohexanone 99+% ACS
Cyclohexanone
Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6
Melting Pt: -31
Density: 0.947 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
UN: 1915
ADR: 3 III
Merck Index: 13,02755

Order Now

Specification Test Results

Appearance Clear viscous liquid
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR