Copper(II) oxide, powder ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC40586-0010
AC40586-0250
AC40586-2500
TS40586-0010EA
574
USD
TS40586-0010
TS40586-0250
TS40586-2500
Copper(II) oxide, powder ACS
Copper (II) oxide
Formula:
CuO Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 1326 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9,III Merck Index: 13,02674 |
Specification Test Results
Appearance | Black to brown-black Powder |
Titration Complexometric | ≥99.0 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤200 ppm |
Calcium (Ca) | ≤100 ppm |
Iron (Fe) | ≤500 ppm |
Potassium (K) | ≤200 ppm |
Sodium (Na) | ≤500 ppm |
Carbon compounds (as C) | ≤0.01 % |
Insoluble matter | ≤0.02 % (in dilute HCl) |
Nitrogen compounds (as N) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...