Copper(II) nitrate hemipentahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate

AC40585-0025 AC40585-1000 AC40585-5000
TS40585-0025EA 250 USD
TS40585-0025 TS40585-1000 TS40585-5000
Copper(II) nitrate hemipentahydrate 98+% ACS
Copper (II) nitrate hemipentahydrate
Formula: Cu(NO₃)₂·2.5H₂O
Melting Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Blue Crystalline powder
Titration Iodometric 98.0 to 102.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤10 ppm
Nickel (Ni) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR