Crystal violet, certified

Supplier: Thermo Scientific Chemicals
Danger

40583-0250 40583-1000 40583-5000
TS40583-0250EA 39 USD
TS40583-0250 TS40583-1000 TS40583-5000
Crystal violet, certified
Crystal Violet
Formula: C₂₅H₃₀ClN₃
Melting Pt: 173 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥88 %
UV Lambda max 589 to 594 nm
Absorption Ratio P-15/P+15 0.98 to 1.20
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR