Hexaamminecobalt trichloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride
AC40531-0050
AC40531-0250
AC40531-1000
TS40531-0050EA
37
USD
TS40531-0050
TS40531-0250
TS40531-1000
Hexaamminecobalt trichloride 99%
Hexaamminecobalt trichloride
Formula:
H₁₈CoCl₃N₆ Melting Pt: 217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036304 CAS Number: 10534-89-1 EINECS: 234-103-9 Merck Index: 12,04711 |
Specification Test Results
Appearance | Orange to orange-brown Powder |
Titration Argentometric | ≥98.5 % |
Refractive index | 1.4170 to 1.4240 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...