2-Chloronicotinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid

40486-0500
TS40486-0500EA 79 USD
TS40486-0500
2-Chloronicotinic acid 99%
2-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤0.3 % (105°C, 3 hrs)
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR