1-Chlorobutane, Sequenation Grade

Supplier: Thermo Scientific Chemicals
Danger

404550010
TS404550010EA 86 USD
TS404550010
1-Chlorobutane, Sequenation Grade
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
Boiling Pt: 77…78 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0.88 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 %
GC ≥99.5 %
UV at 300 nm A: ≤0.005
Residue after evaporation ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR