Calcium hydroxide 95+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Slaked lime
40385-0010
40385-0025
TS40385-0010EA
106
USD
TS40385-0010
TS40385-0025
Calcium hydroxide 95+% ACS
Calcium hydroxide
Formula:
Ca(OH)₂ Boiling Pt: 2850 °C Melting Pt: 580 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 EINECS: 215-137-3 Merck Index: 13,01675 |
Specification Test Results
Appearance | White Powder |
Titration with HCl | ≤3.0 % (CaCO3) |
Chloride (Cl) | ≤300 ppm |
Iron (Fe) | ≤500 ppm |
Magnesium (Mg) | ≤5000 ppm |
Potassium (K) | ≤500 ppm |
Sodium (Na) | ≤500 ppm |
Strontium (Sr) | ≤500 ppm |
Heavy metals (ICP-OES) | ≤30 ppm |
Sulfur compounds (as SO4) | ≤0.1 % |
Insoluble matter | ≤0.03 % (in HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...