1-Butanesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Butanesulphonylchloride, 1-Butanesulfonylchloride

40332-0250 40332-1000
TS40332-0250EA 95 USD
TS40332-0250 TS40332-1000
1-Butanesulphonyl chloride 98%
1-Butanesulphonyl chloride
Formula: C₄H₉ClO₂S
MW: 156.63 g/mol
Boiling Pt: 208
Melting Pt: -25
Density: 1.2 g/cm³
Flash Pt: 79 °C
MDL Number: MFCD00007464
CAS Number: 2386-60-9
EINECS: 219-204-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR