Bromine 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

40284-5000
TS40284-5000EA 235 USD
TS40284-5000
Bromine 99+%, extra pure
Bromine
Formula: Br₂
MW: 159.81 g/mol
Boiling Pt: 58.7 °C (1013 hPa)
Melting Pt: –7.2 °C
Density: 3.111 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010896
CAS Number: 7726-95-6
EINECS: 231-778-1
UN: 1744
ADR: 8,I
Merck Index: 13,01378

Order Now

Specification Test Results

Appearance Red-brown Liquid
Titration Iodometric ≥99.0 %
Chlorine (Cl) ≤500 ppm
Iodine (I) ≤10 ppm
Heavy metals (as Pb) ≤2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR