N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 1,5,9,13-Tetraazatridecane

40228-0250 40228-0050
TS40228-0250EA 568 USD
TS40228-0250 TS40228-0050
N,N'-Bis(3-aminopropyl)propane-1,3-diamine 97%
N,N'-Bis(3-aminopropyl)propane-1,3-diamine
Formula: C₉H₂₄N₄
Boiling Pt: 102 °C (1 mmHg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008213
CAS Number: 4605-14-5
EINECS: 225-007-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4930 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR