Benzoyl chloride 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzoic acid chloride

40203-0025 40203-0050 40203-5000
TS40203-0025EA 532 USD
TS40203-0025 TS40203-0050 TS40203-5000
Benzoyl chloride 98+% ACS
Benzoyl chloride
Formula: C₇H₅ClO
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.21 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II
Merck Index: 13,01113

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20°C, 589 nm)
Residue after ignition ≤0.005 %
Titration excess NaOH 98.0 to 100.5 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Phosphorous compounds (as P) ≤0.002 %
Freezing point -2.0 to 0.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR