Ammonium carbonate ACS

Supplier: Thermo Scientific Chemicals
Warning

AC40113-0010 AC40113-0025 AC40113-0250
TS40113-0010EA 150 USD
TS40113-0010 TS40113-0025 TS40113-0250
Ammonium carbonate ACS
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: 58 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR