(R)-(+)-1,2-Epoxypropane 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-(+)-Propene oxide, (R)-(+)-Methyloxirane, (R)-(+)-1,2-Propylene oxide, (R)-(+)-Propylene oxide

39957-0050 39957-0250
TS39957-0050EA 85 USD
TS39957-0050 TS39957-0250
(R)-(+)-1,2-Epoxypropane 98+%
(R)-(+)-1,2-Epoxypropane
Formula: C₃H₆O
MW: 58.08 g/mol
Melting Pt: –112 °C
Density: 0.829 g/cm³
Flash Pt: –37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066211
CAS Number: 15448-47-2
UN: 1280
ADR: 3 I

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3680 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation +12° to +15° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR