6-Chlorooxindole 98%

Supplier: Thermo Scientific Chemicals
Warning

39944-0250
TS39944-0250EA 148 USD
TS39944-0250
6-Chlorooxindole 98%
6-Chlorooxindole
Formula: C₈H₆ClNO
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00209962
CAS Number: 56341-37-8

Order Now

Specification Test Results

Appearance Off-white to tan Crystalline powder
Infrared spectrum Conforms
Melting point 195 to 199 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR