2-Bromo-5-fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

39797-0050
TS39797-0050EA 113 USD
TS39797-0050
2-Bromo-5-fluoropyridine 98%
2-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 83 °C (44 mmHg)
Melting Pt: 30…31 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless white to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR