Dimethyl sulfoxide 99.8+% for HPLC

Supplier: Thermo Scientific Chemicals

397600010 397600025
TS397600010EA 152 USD
TS397600010 TS397600025
Dimethyl sulfoxide 99.8+% for HPLC
Dimethyl sulphoxide
Formula: (CH₃)₂SO
Boiling Pt: 189 °C (760 mmHg)
Melting Pt: 18.4 °C
Density: 1.1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 %
GC ≥99.80 %
UV at 400 nm A: ≤0.009
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR