Pyridine for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Azabenzene, Azine

396800010
TS396800010EA 255 USD
TS396800010
Pyridine for HPLC
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.978 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
UN: 1282
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.8 %
UV at 400 nm A: ≤0.01
Color scale ≤20 APHA
Residue after evaporation ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR