(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

39662-1000 396620025 396620010
TS39662-1000EA 134 USD
TS39662-1000 TS396620025 TS396620010
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal™
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 79
Melting Pt: -136
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
UN: 2536
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.002 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm (BHT)
Peroxides ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR