1,1'-[Oxybis(ethyleneoxy)]diethylene 95%

Supplier: Thermo Scientific Chemicals

39541-0500
TS39541-0500EA 188 USD
TS39541-0500
1,1'-[Oxybis(ethyleneoxy)]diethylene 95%
1,1'-[Oxybis(ethyleneoxy)]diethylene
Formula: H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: –21 °C
Density: 0.97 g/cm³
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054610
CAS Number: 764-99-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR