Phenyl propargyl ether 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Prop-2-ynyloxy)benzene, (Prop-2-yn-1-yloxy)benzene

39273-0050 39273-0250
TS39273-0050EA 74 USD
TS39273-0050 TS39273-0250
Phenyl propargyl ether 97+%
Phenyl propargyl ether
Formula: C₉H₈O
Boiling Pt: 89…90 °C (14 mmHg)
Density: 1.03 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048107
CAS Number: 13610-02-1
EINECS: 237-095-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20°C, 589 nm)
GC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR