5-Methylsalicylaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

39263-0010 39263-0050
TS39263-0010EA 39 USD
TS39263-0010 TS39263-0050
5-Methylsalicylaldehyde 99%
5-Methylsalicylaldehyde
Formula: C₈H₈O₂
Boiling Pt: 217 °C (760 mmHg)
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00274297
CAS Number: 613-84-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR