Benzyl alcohol for analysis

Supplier: Thermo Scientific Chemicals
Warning

39122-0025 39122-0010
TS39122-0025EA 262 USD
TS39122-0025 TS39122-0010
Benzyl alcohol for analysis
Benzyl alcohol
Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.044 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.1 % (K.F.)
GC ≥99.5 %
Color scale ≤20 APHA
Acidity ≤0.001 meq/g
Alkalinity ≤0.002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Halogenated compounds ≤0.001 % (as Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR