N,N-Dimethylacetamide 99.5+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

390800010
TS390800010EA 104 USD
TS390800010
N,N-Dimethylacetamide 99.5+% for analysis
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.937 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.50 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.001 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR