tert-Butyl methyl ether for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MTBE, 2-Methyl-2-methoxypropane, Ether, tert-butyl methyl, Methyl 1,1-dimethylethyl ether, 2-Methoxy-2-methylpropane, Methyl t-butyl ether, Methyl tert-butyl ether, Methyl-t-Butyl Ether

390480025 390480010
TS390480025EA 133 USD
TS390480025 TS390480010
tert-Butyl methyl ether for analysis
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2
Melting Pt: -108.6
Density: 0.7404 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3 II
Merck Index: 13,06059

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.5 %
Residue after evaporation ≤0.005 %
Acidity ≤0.0005 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Boron (B) ≤0.02 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR