3-Chloro-4-methylphenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

390430050
TS390430050EA 57 USD
TS390430050
3-Chloro-4-methylphenyl isocyanate 98%
3-Chloro-4-methylphenyl isocyanate
Formula: C₈H₆ClNO
Boiling Pt: 225 °C (760 mmHg)
Melting Pt: 23 °C
Density: 1.23 g/cm³
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013858
CAS Number: 28479-22-3
UN: 3428
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5580 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR