Calcium citrate tetrahydrate 98+% for analysis
Supplier: Thermo Scientific Chemicals
AC39008-0010
AC39008-2500
TS39008-0010EA
87
USD
TS39008-0010
TS39008-2500
Calcium citrate tetrahydrate 98+% for analysis
Calcium citrate tetrahydrate
Formula:
C₁₂H₁₀Ca₃O₁₄·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150786 CAS Number: 5785-44-4 EINECS: 212-391-7 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Complexometric | 98.0 to 102.0 % (on dried basis) |
Loss on drying | 10.0 % to 13.3 % (150°C, 4 h) |
Fluoride (F) | ≤0.003 % |
Arsenic (As) | ≤0.0003 % |
Lead (Pb) | ≤0.001 % |
Heavy metals (as Pb) | ≤0.002 % |
Insoluble matter | ≤0.2 % (in HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...