Calcium citrate tetrahydrate 98+% for analysis

Supplier: Thermo Scientific Chemicals

AC39008-0010 AC39008-2500
TS39008-0010EA 87 USD
TS39008-0010 TS39008-2500
Calcium citrate tetrahydrate 98+% for analysis
Calcium citrate tetrahydrate
Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Complexometric 98.0 to 102.0 % (on dried basis)
Loss on drying 10.0 % to 13.3 % (150°C, 4 h)
Fluoride (F) ≤0.003 %
Arsenic (As) ≤0.0003 %
Lead (Pb) ≤0.001 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.2 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR