N-Boc-ethylenediamine 98%

Supplier: Thermo Scientific Chemicals

38990-0010 38990-0050 38990-0250
TS38990-0010EA 87 USD
TS38990-0010 TS38990-0050 TS38990-0250
N-Boc-ethylenediamine 98%
N-Boc-ethylenediamine

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR