1-Propanol for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

389600010 389600025
TS389600010EA 132 USD
TS389600010 TS389600025
1-Propanol for HPLC
1-Propanol
Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: -127
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
UN: 1274
ADR: 3 II
Merck Index: 13,07934

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.9 %
UV at 250 nm A: ≤0.05
Non-volatile matter ≤0.0003 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR