Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium tetrathiomolybdate(VI)
AC38953-0010
AC38953-0100
AC38953-0500
AC38953-2500
TS38953-0010EA
45
USD
TS38953-0010
TS38953-0100
TS38953-0500
TS38953-2500
Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Ammonium tetrathiomolybdate
Formula:
H₁₀MoN₂S₄ Melting Pt: 300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00136013 CAS Number: 15060-55-6 |
Specification Test Results
Appearance | Brown to black Powder or crystals |
Total trace metal impurities | ≤500 ppm |
Molybdenum (Mo) | 35.5 to 38.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...