Ammonium tetrathiomolybdate 99.95% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium tetrathiomolybdate(VI)

AC38953-0010 AC38953-0100 AC38953-0500 AC38953-2500
TS38953-0010EA 45 USD
TS38953-0010 TS38953-0100 TS38953-0500 TS38953-2500
Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Ammonium tetrathiomolybdate
Formula: H₁₀MoN₂S₄
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00136013
CAS Number: 15060-55-6

Order Now

Specification Test Results

Appearance Brown to black Powder or crystals
Total trace metal impurities ≤500 ppm
Molybdenum (Mo) 35.5 to 38.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR