Zinc chloride 0.7 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

389451000 389458000
TS389451000EA 131 USD
TS389451000 TS389458000
Zinc chloride 0.7 M in THF, AcroSeal™
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 66 °C
Flash Pt: –22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear to hazy colorless Solution
Titration Argentometric 0.65 to 0.8 M
Water ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR