Water for analysis

Supplier: Thermo Scientific Chemicals

389400025 389400100 389400010
TS389400025EA 50 USD
TS389400025 TS389400100 TS389400010
Water for analysis
Water
Formula: H₂O
Boiling Pt: 100 °C (760 mmHg)
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Non-volatile matter ≤0.0005 %
Residue after ign. (sulfated) ≤0.0002 %
pH 5 to 8
Absorbance ≤0.01 (water, 1 cm, 254 nm)
Chloride (Cl) ≤0.1 ppm
Nitrate (NO3) ≤0.2 ppm
Phosphate (PO4) ≤0.05 ppm
Sulfate (SO4) ≤1 ppm
Silicate (as SiO2) ≤0.01 ppm
Aluminium (Al) ≤0.02 ppm
Ammonium (NH4) ≤0.01 ppm
Antimony (Sb) ≤0.01 ppm
Barium (Ba) ≤0.01 ppm
Bismuth (Bi) ≤0.01 ppm
Boron (B) ≤0.01 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.1 ppm
Chromium (Cr) ≤0.01 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.01 ppm
Gold (Au) ≤0.01 ppm
Iron (Fe) ≤0.01 ppm
Lithium (Li) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Manganese (Mn) ≤0.01 ppm
Mercury (Hg) ≤0.05 ppm
Molybdenum (Mo) ≤0.01 ppm
Nickel (Ni) ≤0.01 ppm
Potassium (K) ≤0.05 ppm
Selenium (Se) ≤0.01 ppm
Silver (Ag) ≤0.01 ppm
Sodium (Na) ≤0.1 ppm
Strontium (Sr) ≤0.05 ppm
Tin (Sn) ≤0.01 ppm
Titanium (Ti) ≤0.01 ppm
Vanadium (V) ≤0.01 ppm
Zinc (Zn) ≤0.05 ppm
Zirconium (Zr) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity ≤6 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR