Soda lime, granular, pure indicator, with indicator

Supplier: Thermo Scientific Chemicals
Danger

388350010 388350500 388352500
TS388350010EA 110 USD
TS388350010 TS388350500 TS388352500
Soda lime, granular, pure indicator, with indicator
Soda lime

With indicator white to violet

Storage Temperature: Ambient
MDL Number: MFCD00134094
CAS Number: 8006-28-8
Merck Index: 13,08640

Order Now

Specification Test Results

Appearance White to beige to light grey Granules
Loss on drying ≤20 % (10 g, 200°C, 2 h)
CO2 absorption capacity ≥19.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR