Acetyl chloride 1 M in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetyl chloride 1M in Dichloromethane

387741000 387748000
TS387741000EA 53 USD
TS387741000 TS387748000
Acetyl chloride 1 M in dichloromethane, AcroSeal®
Acetyl chloride
Formula: C₂H₃ClO
Storage Temperature: Ambient
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Argentometric 0.9 to 1.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR