Citric acid monohydrate 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

38585-0010 38585-0025 38585-2500
TS38585-0010EA 68 USD
TS38585-0010 TS38585-0025 TS38585-2500
Citric acid monohydrate 99+% ACS
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals
Residue after ignition ≤0.02 %
Titration with NaOH 99.0 to 102.0 %
Carboniz. sub. by hot H2SO4 Passes test
Chloride (Cl) ≤0.001 %
Oxalate (C2O4) (limit about 0.05%) Passes test
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤3 ppm
Lead (Pb) ≤2 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR