Sodium dichromate dihydrate 99.5-100.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

AC38563-0025 AC38563-1000 AC38563-5000
TS38563-0025EA 431 USD
TS38563-0025 TS38563-1000 TS38563-5000
Sodium dichromate dihydrate 99.5-100.5% ACS
Sodium dichromate dihydrate
Formula: Na₂Cr₂O₇·2H₂O
Boiling Pt: 400 °C (760 mmHg)
Melting Pt: 357 °C
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
UN: 3087
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Titration Iodometric 99.5 to 100.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.01 %
Aluminium (Al) ≤0.002 %
Calcium (Ca) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR