Sodium hydroxide 50% in water for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

380210025 380210100 380215000
TS380210025EA 128 USD
TS380210025 TS380210100 TS380215000
Sodium hydroxide 50% in water for analysis
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Viscous solution
Titration with HCl 49 to 51 %
Sodium carbonate (Na2CO3) ≤0.1 %
Sodium chloride (NaCl) ≤0.008 %
Iron (Fe) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR