Sodium hydroxide 50% in water for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
380210025
380210100
380215000
TS380210025EA
128
USD
TS380210025
TS380210100
TS380215000
Sodium hydroxide 50% in water for analysis
Sodium hydroxide
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 12 °C Density: 1.5 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Viscous solution |
Titration with HCl | 49 to 51 % |
Sodium carbonate (Na2CO3) | ≤0.1 % |
Sodium chloride (NaCl) | ≤0.008 % |
Iron (Fe) | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...