2,5-Dichloro-3-nitropyridine 97%

Warning

37952-0010
TS37952-0010EA 28 USD
TS37952-0010 TS37952-0050
2,5-Dichloro-3-nitropyridine 97%
2,5-Dichloro-3-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Melting Pt: 39…45 °C
Storage Temperature: Ambient
MDL Number: MFCD06658963
CAS Number: 21427-62-3

Order Now

Specification Test Results

Appearance Green-beige to orange Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 39 to 45 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR