Lithium-tert-butanolate 2.2 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

37931-1000 AC37931-8000
TS37931-1000EA 115 USD
TS37931-1000 TS37931-8000
Lithium-tert-butanolate 2.2 M in THF, AcroSeal™
Lithium tert-butanolate
Formula: C₄H₉LiO
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00050479
CAS Number: 1907-33-1
EINECS: 217-611-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Solution
Strength 19 to 21 %
Impurity ≤0.2 % LiOH + Li2CO3 (as LiOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR