Lithium hydroxide, anhydrous 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

AC37918-0025 AC37918-1000 AC37918-5000
TS37918-0025EA 931 USD
TS37918-0025 TS37918-1000 TS37918-5000
Lithium hydroxide, anhydrous 98%, pure
Lithium hydroxide
Formula: LiOH
Melting Pt: 462 °C
Density: 1.43 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011095
CAS Number: 1310-65-2
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Titration with HCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR